Product Details

SNP ID
rs78399510
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:203482129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCTTTCTGGATTTAACCAGGA[C/T]GCTAAGGTTCATACATCCTAACATC
Phenotype
MIM: 601914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRELP PubMed Links

Gene Details

Gene
PRELP
Gene Name
proline and arginine rich end leucine rich repeat protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002725.3 Intron NP_002716.1
NM_201348.1 Intron NP_958505.1

View Full Product Details