Product Details

SNP ID
rs78773986
Assay Type
Functionally Tested
NCBI dbSNP Submissions
18
Location
Chr.1:204032792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAAGGAAGGAAACTTCAAGGAG[A/G]CCCTTAGCCTTTACAGTTGAACTGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details