Product Details

SNP ID
rs77568723
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:155135677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGGACTGACCGGGACATGGA[A/G]GAGGTCTGGACCAGGGTACTGGGAA
Phenotype
MIM: 605951 MIM: 191164 MIM: 613683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM3 PubMed Links

Gene Details

Gene
DPM3
Gene Name
dolichyl-phosphate mannosyltransferase subunit 3
There are no transcripts associated with this gene.

Gene
EFNA1
Gene Name
ephrin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004428.2 424 Intron NP_004419.2
NM_182685.1 424 Intron NP_872626.1
Gene
SLC50A1
Gene Name
solute carrier family 50 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122837.1 424 Intron NP_001116309.1
NM_001122839.1 424 Intron NP_001116311.1
NM_001287586.1 424 Intron NP_001274515.1
NM_001287587.1 424 Intron NP_001274516.1
NM_001287588.1 424 Intron NP_001274517.1
NM_001287589.1 424 Intron NP_001274518.1
NM_001287590.1 424 Silent Mutation GAA,GAG E2E NP_001274519.1
NM_001287591.1 424 Intron NP_001274520.1
NM_001287592.1 424 Intron NP_001274521.1
NM_018845.3 424 Intron NP_061333.2
XM_005245339.4 424 Intron XP_005245396.1
XM_005245340.3 424 Silent Mutation GAA,GAG E2E XP_005245397.1
XM_005245341.4 424 UTR 5 XP_005245398.1
XM_005245342.3 424 Intron XP_005245399.1
XM_006711453.2 424 Intron XP_006711516.1

View Full Product Details