Product Details

SNP ID
rs76114442
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:156730185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCACGACAATCCCTTGCTTCCTA[A/G]ATACTTTTATGAAATCCTAACTCCT
Phenotype
MIM: 611930 MIM: 611836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISG20L2 PubMed Links

Gene Details

Gene
ISG20L2
Gene Name
interferon stimulated exonuclease gene 20 like 2
There are no transcripts associated with this gene.

Gene
MRPL24
Gene Name
mitochondrial ribosomal protein L24
There are no transcripts associated with this gene.

Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142560.1 Intron NP_001136032.1
NM_015997.3 Intron NP_057081.3
XM_005245234.4 Intron XP_005245291.1
XM_005245235.4 Intron XP_005245292.1
XM_005245236.4 Intron XP_005245293.1
XM_005245237.4 Intron XP_005245294.1
XM_011509600.1 Intron XP_011507902.1
XM_017001404.1 Intron XP_016856893.1
XM_017001405.1 Intron XP_016856894.1
XM_017001406.1 Intron XP_016856895.1

View Full Product Details