Product Details
- SNP ID
-
rs75965541
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:157830210 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTCACATTTCATATCTGAATAAT[G/T]CAGGGTAAACATGACAAGGATGCCC
- Phenotype
-
MIM: 602592
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CD5L
PubMed Links
Gene Details
- Gene
- CD5L
- Gene Name
- CD5 molecule like
View Full Product Details