Product Details

SNP ID
rs78302998
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:26442637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGGAGGAGAAAAGTCTGTCTC[C/T]TATCTCCAACTCTGAGTCAGGACCA
Phenotype
MIM: 608172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHDDS PubMed Links

Gene Details

Gene
DHDDS
Gene Name
dehydrodolichyl diphosphate synthase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243564.1 Intron NP_001230493.1
NM_001243565.1 Intron NP_001230494.1
NM_001319959.1 Intron NP_001306888.1
NM_024887.3 Intron NP_079163.2
NM_205861.2 Intron NP_995583.1
XM_006710912.2 Intron XP_006710975.1
XM_006710913.2 Intron XP_006710976.1
XM_006710914.2 Intron XP_006710977.1
XM_006710915.2 Intron XP_006710978.1
XM_006710916.2 Intron XP_006710979.1
XM_006710917.2 Intron XP_006710980.1
XM_006710918.2 Intron XP_006710981.1
XM_011542183.2 Intron XP_011540485.1
XM_011542184.2 Intron XP_011540486.1
XM_011542185.2 Intron XP_011540487.1
XM_011542186.2 Intron XP_011540488.1
XM_017002379.1 Intron XP_016857868.1
XM_017002380.1 Intron XP_016857869.1
XM_017002381.1 Intron XP_016857870.1
XM_017002382.1 Intron XP_016857871.1
XM_017002383.1 Intron XP_016857872.1
XM_017002384.1 Intron XP_016857873.1

View Full Product Details