Product Details

SNP ID
rs75735055
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:29120795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCTGTGGTCAGAGCATCCATC[A/C]CCAGCCTGGGATGTGACTGAAACAT
Phenotype
MIM: 130500
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166005.1 1108 Intron NP_001159477.1
NM_001166006.1 1108 Intron NP_001159478.1
NM_001166007.1 1108 Intron NP_001159479.1
NM_004437.3 1108 Intron NP_004428.1
NM_203342.2 1108 Intron NP_976217.1
NM_203343.2 1108 Intron NP_976218.1
XM_005245753.1 1108 Intron XP_005245810.1
XM_005245757.1 1108 Intron XP_005245814.1
XM_005245760.1 1108 Intron XP_005245817.1
XM_005245761.1 1108 Intron XP_005245818.1
XM_005245763.1 1108 Intron XP_005245820.1
XM_005245764.1 1108 Intron XP_005245821.1
XM_005245765.1 1108 Intron XP_005245822.1
XM_005245768.1 1108 Intron XP_005245825.1
XM_005245769.1 1108 Intron XP_005245826.1
XM_005245770.1 1108 Intron XP_005245827.1
XM_005245772.4 1108 Intron XP_005245829.1
XM_005245773.4 1108 Intron XP_005245830.1
XM_005245774.1 1108 Intron XP_005245831.1
XM_006710434.1 1108 Intron XP_006710497.1
XM_006710439.1 1108 Intron XP_006710502.1
XM_011540956.1 1108 Intron XP_011539258.1
XM_011540957.1 1108 Intron XP_011539259.1
XM_011540958.1 1108 Intron XP_011539260.1
XM_011540959.1 1108 Intron XP_011539261.1
XM_011540960.1 1108 Intron XP_011539262.1
XM_011540961.1 1108 Intron XP_011539263.1
XM_011540962.1 1108 Intron XP_011539264.1
XM_011540963.2 1108 Intron XP_011539265.1
XM_011540964.1 1108 Intron XP_011539266.1
XM_011540965.2 1108 Intron XP_011539267.1
XM_017000581.1 1108 Intron XP_016856070.1
XM_017000582.1 1108 Intron XP_016856071.1
XM_017000583.1 1108 Intron XP_016856072.1
XM_017000584.1 1108 Intron XP_016856073.1
XM_017000585.1 1108 Intron XP_016856074.1
XM_017000586.1 1108 Intron XP_016856075.1
XM_017000587.1 1108 Intron XP_016856076.1
XM_017000588.1 1108 Intron XP_016856077.1
XM_017000589.1 1108 Intron XP_016856078.1
XM_017000590.1 1108 Intron XP_016856079.1
XM_017000591.1 1108 Intron XP_016856080.1
XM_017000592.1 1108 Intron XP_016856081.1
XM_017000593.1 1108 Intron XP_016856082.1
XM_017000594.1 1108 Intron XP_016856083.1
XM_017000595.1 1108 Intron XP_016856084.1
XM_017000596.1 1108 Intron XP_016856085.1
XM_017000597.1 1108 Intron XP_016856086.1
XM_017000598.1 1108 Intron XP_016856087.1
XM_017000599.1 1108 Intron XP_016856088.1
XM_017000600.1 1108 Intron XP_016856089.1
XM_017000601.1 1108 Intron XP_016856090.1
XM_017000602.1 1108 Intron XP_016856091.1
XM_017000603.1 1108 Intron XP_016856092.1
XM_017000604.1 1108 Intron XP_016856093.1
Gene
TMEM200B
Gene Name
transmembrane protein 200B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003682.3 1108 UTR 3 NP_001003682.1
NM_001171868.1 1108 UTR 3 NP_001165339.1
XM_011541457.2 1108 UTR 3 XP_011539759.1

View Full Product Details