Product Details

SNP ID
rs77796699
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:16443885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTTTAAGCTGTACCATTCCCC[C/T]ACCTTAGTTGTAGAAAGTACCGTCT
Phenotype
MIM: 611624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NECAP2 PubMed Links

Gene Details

Gene
NECAP2
Gene Name
NECAP endocytosis associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145277.1 Intron NP_001138749.1
NM_001145278.1 Intron NP_001138750.1
NM_018090.4 Intron NP_060560.1
XM_011541767.2 Intron XP_011540069.1
XM_017001721.1 Intron XP_016857210.1
Gene
SPATA21
Gene Name
spermatogenesis associated 21
There are no transcripts associated with this gene.

View Full Product Details