Product Details

SNP ID
rs79080076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:15594877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCTCTGGTCTTCATATAGCCCT[A/G]CAAGGTAGGCAATATCAGCCCCATT
Phenotype
MIM: 601257
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX1 PubMed Links

Gene Details

Gene
DDX1
Gene Name
DEAD/H-box helicase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004939.2 Intron NP_004930.1

View Full Product Details