Product Details

SNP ID
rs74768261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8731967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTGAGACTCGAAAGACTTGGGG[C/T]TCTGCGAGTTTGTGACTGTGAAGAC
Phenotype
MIM: 615759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIDINS220 PubMed Links

Gene Details

Gene
KIDINS220
Gene Name
kinase D-interacting substrate 220kDa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020738.2 4251 Missense Mutation ACC,GCC T1357A NP_065789.1

View Full Product Details