Product Details

SNP ID
rs79638552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9480618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGACCGGAAAGATATACATCC[A/C]AATGCCAACAGTGGTTTTCTGGGTG
Phenotype
MIM: 603639 MIM: 606029
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAM17 PubMed Links

Gene Details

Gene
ADAM17
Gene Name
ADAM metallopeptidase domain 17
There are no transcripts associated with this gene.

Gene
CPSF3
Gene Name
cleavage and polyadenylation specific factor 3
There are no transcripts associated with this gene.

Gene
IAH1
Gene Name
isoamyl acetate-hydrolyzing esterase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039613.2 Intron NP_001034702.1
NM_001320858.1 Intron NP_001307787.1
NM_001320859.1 Intron NP_001307788.1
NM_001320860.1 Intron NP_001307789.1
NM_001320863.1 Intron NP_001307792.1
XM_017003878.1 Intron XP_016859367.1

View Full Product Details