Product Details

SNP ID
rs80212211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151487304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGTATATAATAAGTGATAATA[C/T]CAAAAGTTTCTTCCTAGGAGCAGCC
Phenotype
MIM: 161650 MIM: 608952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEB PubMed Links

Gene Details

Gene
NEB
Gene Name
nebulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164507.1 Intron NP_001157979.1
NM_001164508.1 Intron NP_001157980.1
NM_001271208.1 Intron NP_001258137.1
NM_004543.4 Intron NP_004534.2
XM_005246590.2 Intron XP_005246647.1
XM_005246591.2 Intron XP_005246648.1
XM_005246592.2 Intron XP_005246649.1
XM_005246593.2 Intron XP_005246650.1
XM_005246594.2 Intron XP_005246651.1
XM_005246596.2 Intron XP_005246653.1
XM_005246597.2 Intron XP_005246654.1
XM_005246598.2 Intron XP_005246655.1
XM_005246599.2 Intron XP_005246656.1
XM_005246600.2 Intron XP_005246657.1
XM_005246601.2 Intron XP_005246658.1
XM_005246602.2 Intron XP_005246659.1
XM_005246603.2 Intron XP_005246660.1
XM_005246604.2 Intron XP_005246661.1
XM_005246606.2 Intron XP_005246663.1
XM_005246608.2 Intron XP_005246665.1
XM_005246610.2 Intron XP_005246667.1
XM_005246611.2 Intron XP_005246668.1
XM_005246612.2 Intron XP_005246669.1
XM_005246613.2 Intron XP_005246670.1
XM_005246615.2 Intron XP_005246672.1
XM_005246616.1 Intron XP_005246673.1
XM_005246617.2 Intron XP_005246674.1
XM_006712541.2 Intron XP_006712604.1
XM_006712542.2 Intron XP_006712605.1
XM_011511225.2 Intron XP_011509527.1
XM_011511226.2 Intron XP_011509528.1
XM_011511227.2 Intron XP_011509529.1
XM_017004177.1 Intron XP_016859666.1
XM_017004178.1 Intron XP_016859667.1
XM_017004179.1 Intron XP_016859668.1
XM_017004180.1 Intron XP_016859669.1
XM_017004181.1 Intron XP_016859670.1
XM_017004182.1 Intron XP_016859671.1
XM_017004183.1 Intron XP_016859672.1
XM_017004184.1 Intron XP_016859673.1
XM_017004185.1 Intron XP_016859674.1
Gene
RIF1
Gene Name
replication timing regulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177663.1 Intron NP_001171134.1
NM_001177664.1 Intron NP_001171135.1
NM_001177665.1 Intron NP_001171136.1
NM_018151.4 Intron NP_060621.3
XM_005246665.3 Intron XP_005246722.1
XM_011511395.2 Intron XP_011509697.1
XM_017004422.1 Intron XP_016859911.1
XM_017004423.1 Intron XP_016859912.1
XM_017004424.1 Intron XP_016859913.1
XM_017004425.1 Intron XP_016859914.1

View Full Product Details