Product Details

SNP ID
rs80320708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202940619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTAGCTGCATTTTTTTCTTACC[A/G]GAAGCCAGGTTACACTCTTCACCCA
Phenotype
MIM: 607586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARF PubMed Links

Gene Details

Gene
CARF
Gene Name
calcium responsive transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104586.2 Intron NP_001098056.1
NM_001282910.1 Intron NP_001269839.1
NM_001282911.1 Intron NP_001269840.1
NM_001282912.1 Intron NP_001269841.1
NM_001322427.1 Intron NP_001309356.1
NM_001322428.1 Intron NP_001309357.1
NM_001322429.1 Intron NP_001309358.1
NM_001322430.1 Intron NP_001309359.1
NM_024744.15 Intron NP_079020.13
XM_005246858.3 Intron XP_005246915.1
XM_005246859.3 Intron XP_005246916.1
XM_005246864.3 Intron XP_005246921.1
XM_006712760.2 Intron XP_006712823.1
XM_011511867.2 Intron XP_011510169.1
XM_011511868.2 Intron XP_011510170.1
XM_011511869.2 Intron XP_011510171.1
XM_017004961.1 Intron XP_016860450.1
XM_017004962.1 Intron XP_016860451.1
XM_017004963.1 Intron XP_016860452.1
XM_017004964.1 Intron XP_016860453.1
XM_017004965.1 Intron XP_016860454.1
XM_017004966.1 Intron XP_016860455.1

View Full Product Details