Product Details

SNP ID
rs380460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60146245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAACCTAGATGTAAAAAAAATG[C/T]GAGATTTGCTAAATGATGTTTATAT
Phenotype
MIM: 612833
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS7 PubMed Links

Gene Details

Gene
DHRS7
Gene Name
dehydrogenase/reductase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322280.1 Intron NP_001309209.1
NM_001322281.1 Intron NP_001309210.1
NM_001322282.1 Intron NP_001309211.1
NM_016029.3 Intron NP_057113.1
XM_017021368.1 Intron XP_016876857.1
Gene
PCNX4
Gene Name
pecanex homolog 4 (Drosophila)
There are no transcripts associated with this gene.

View Full Product Details