Product Details

SNP ID
rs571257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49729541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACTAAATTCATCATCCTATCTA[A/C]AAGCTTGACATTTCCTTATTATTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRISP3 PubMed Links

Gene Details

Gene
CRISP3
Gene Name
cysteine rich secretory protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190986.1 Intron NP_001177915.1
NM_006061.2 Intron NP_006052.2

View Full Product Details