Product Details
- SNP ID
-
rs340874
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
73
- Location
-
Chr.1:213985913 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTTTTAAGGTGTGGAAAGGTATA[C/T]AGCCCTTACTCCTACCAGTTTATTT
- Phenotype
-
MIM: 601546
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147923136] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PROX1
- Gene Name
- prospero homeobox 1
- Gene
- PROX1-AS1
- Gene Name
- PROX1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details