Product Details

SNP ID
rs397891
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89686623 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTGGTAACCGTTGCTAAGGAGA[C/G]GAAGCCCGGGAAGGAGCTTTCGCGC
Phenotype
MIM: 603464
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 12 Intron NP_001092003.2
NM_001160367.1 12 Intron NP_001153839.1
NM_052987.3 12 Intron NP_443713.2
NM_052988.4 12 Intron NP_443714.3
XM_006721308.2 12 UTR 5 XP_006721371.1
XM_006721310.2 12 UTR 5 XP_006721373.1
XM_011523405.2 12 UTR 5 XP_011521707.1
XM_011523406.2 12 UTR 5 XP_011521708.1
XM_011523407.2 12 UTR 5 XP_011521709.1
XM_011523408.2 12 UTR 5 XP_011521710.1
XM_011523410.2 12 Intron XP_011521712.1
XM_011523411.2 12 Intron XP_011521713.1
XM_011523412.1 12 Intron XP_011521714.1
XM_011523413.2 12 Intron XP_011521715.1
XM_011523414.2 12 Intron XP_011521716.1
XM_011523415.2 12 Intron XP_011521717.1
XM_011523416.2 12 Intron XP_011521718.1
XM_011523417.2 12 UTR 5 XP_011521719.1
XM_017023806.1 12 UTR 5 XP_016879295.1
XM_017023807.1 12 UTR 5 XP_016879296.1
XM_017023808.1 12 Intron XP_016879297.1
XM_017023809.1 12 Intron XP_016879298.1
XM_017023810.1 12 Intron XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

View Full Product Details