Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001024218.1 | Intron | NP_001019389.1 | |||
NM_020806.4 | Intron | NP_065857.1 | |||
XM_005267254.3 | Intron | XP_005267311.1 | |||
XM_011536340.2 | Intron | XP_011534642.1 | |||
XM_011536342.2 | Intron | XP_011534644.1 | |||
XM_011536343.2 | Intron | XP_011534645.1 | |||
XM_011536344.2 | Intron | XP_011534646.1 | |||
XM_011536345.2 | Intron | XP_011534647.1 | |||
XM_011536346.2 | Intron | XP_011534648.1 | |||
XM_011536347.2 | Intron | XP_011534649.1 | |||
XM_017020913.1 | Intron | XP_016876402.1 | |||
XM_017020914.1 | Intron | XP_016876403.1 | |||
XM_017020915.1 | Intron | XP_016876404.1 | |||
XM_017020916.1 | Intron | XP_016876405.1 | |||
XM_017020917.1 | Intron | XP_016876406.1 | |||
XM_017020918.1 | Intron | XP_016876407.1 | |||
XM_017020919.1 | Intron | XP_016876408.1 | |||
XM_017020920.1 | Intron | XP_016876409.1 | |||
XM_017020921.1 | Intron | XP_016876410.1 | |||
XM_017020922.1 | Intron | XP_016876411.1 | |||
XM_017020923.1 | Intron | XP_016876412.1 | |||
XM_017020924.1 | Intron | XP_016876413.1 | |||
XM_017020925.1 | Intron | XP_016876414.1 | |||
XM_017020926.1 | Intron | XP_016876415.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020715.2 | Intron | NP_065766.1 | |||
XM_011537009.1 | Intron | XP_011535311.1 | |||
XM_011537015.2 | Intron | XP_011535317.1 | |||
XM_017021499.1 | Intron | XP_016876988.1 | |||
XM_017021500.1 | Intron | XP_016876989.1 | |||
XM_017021501.1 | Intron | XP_016876990.1 | |||
XM_017021502.1 | Intron | XP_016876991.1 | |||
XM_017021503.1 | Intron | XP_016876992.1 | |||
XM_017021504.1 | Intron | XP_016876993.1 |