Product Details

SNP ID
rs243828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55509211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGTGCGCCCAGGCGGCGGAAGT[G/T]GCGGCCACAGTGCCAGGTGCCGGCG
Phenotype
MIM: 612040 MIM: 120360
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LPCAT2 PubMed Links

Gene Details

Gene
LPCAT2
Gene Name
lysophosphatidylcholine acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017839.4 211 Silent Mutation GTG,GTT V10V NP_060309.2
XM_005256006.2 211 Silent Mutation GTG,GTT V10V XP_005256063.1
XM_011523169.2 211 UTR 5 XP_011521471.1
XM_017023376.1 211 UTR 5 XP_016878865.1
Gene
MMP2
Gene Name
matrix metallopeptidase 2
There are no transcripts associated with this gene.

View Full Product Details