Product Details

SNP ID
rs28940878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:89178078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCTGTCCACCGTGGAGCGGGG[A/G]CAGGAGTCCCTGTGGCCAGCTTTCA
Phenotype
MIM: 606933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TYR PubMed Links

Gene Details

Gene
TYR
Gene Name
tyrosinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000372.4 514 Missense Mutation GAC,GGC D42G NP_000363.1
XM_011542970.2 514 Missense Mutation GAC,GGC D42G XP_011541272.1

View Full Product Details