Product Details

SNP ID
rs555314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42901397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTTTGAGCAAAGAGTTTTATTA[A/G]TATGAAGCAGAAGAGGTAACATAAA
Phenotype
MIM: 613742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G6PC PubMed Links

Gene Details

Gene
G6PC
Gene Name
glucose-6-phosphatase catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000151.3 Intron NP_000142.2
NM_001270397.1 Intron NP_001257326.1
Gene
LINC00671
Gene Name
long intergenic non-protein coding RNA 671
There are no transcripts associated with this gene.

View Full Product Details