Product Details

SNP ID
rs2032594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12701788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTTCCACTGGAGGGAAACAGTT[C/T]AAAGGATATATGTGATACTATTAAT
Phenotype
MIM: 400005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTTY15 PubMed Links

Gene Details

Gene
TTTY15
Gene Name
testis-specific transcript, Y-linked 15 (non-protein coding)
There are no transcripts associated with this gene.

Gene
USP9Y
Gene Name
ubiquitin specific peptidase 9, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004654.3 558 UTR 5 NP_004645.2
XM_017030078.1 558 Intron XP_016885567.1

View Full Product Details