Product Details

SNP ID
rs2338755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39263064 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTATACCTGGGTCTCTCTATTA[C/T]GGTGGATAGGTCCAAAAGCAAAGGT
Phenotype
MIM: 609086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL20 PubMed Links

Gene Details

Gene
FBXL20
Gene Name
F-box and leucine rich repeat protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184906.1 Intron NP_001171835.1
NM_032875.2 Intron NP_116264.2
XM_005257746.3 Intron XP_005257803.2
XM_005257747.3 Intron XP_005257804.2

View Full Product Details