Product Details

SNP ID
rs2271029
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:42683904 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTACTACAGTCTCCTTACTGCGCCG[A/C]GATTCGCCAGGCTGCACGGTGAGCT
Phenotype
MIM: 600240 MIM: 602346
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCR10 PubMed Links

Gene Details

Gene
CCR10
Gene Name
C-C motif chemokine receptor 10
There are no transcripts associated with this gene.

Gene
CNTNAP1
Gene Name
contactin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003632.2 1502 Silent Mutation AGA,CGA R51R NP_003623.1
XM_005257748.4 1502 UTR 5 XP_005257805.1
XM_017025238.1 1502 Silent Mutation AGA,CGA R51R XP_016880727.1
Gene
PLEKHH3
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
There are no transcripts associated with this gene.

View Full Product Details