Product Details

SNP ID
rs1563945
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:28317369 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGACCGCGACTAACTGGACTAACTG[A/C]GTGGAGCTCCGGGAGGGCAGGGCCA
Phenotype
MIM: 601459
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PNOC PubMed Links

Gene Details

Gene
PNOC
Gene Name
prepronociceptin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284244.1 Intron NP_001271173.1
NM_006228.4 Intron NP_006219.1
XM_005273532.2 Intron XP_005273589.1
XM_011544559.2 Intron XP_011542861.1
XM_017013578.1 Intron XP_016869067.1

View Full Product Details