Product Details
- SNP ID
-
rs2878539
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:1250089 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTACTCTGAGCCCTCTGGAATGC[C/T]GAGATAGTTGCATAATTTCAGAAAC
- Phenotype
-
MIM: 602618
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTBP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77350626] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTBP1
- Gene Name
- C-terminal binding protein 1
- Gene
- CTBP1-AS2
- Gene Name
- CTBP1 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.
View Full Product Details