Product Details

SNP ID
rs11871533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35256683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCCAAGTGATTTACTATTACA[C/T]GATGCCACCTTCCTTTTAGTGATTG
Phenotype
MIM: 614952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371932 PubMed Links

Gene Details

Gene
LOC105371932
Gene Name
uncharacterized LOC105371932
There are no transcripts associated with this gene.

Gene
SLFN5
Gene Name
schlafen family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144975.3 Intron NP_659412.3
XM_005257934.3 Intron XP_005257991.1

View Full Product Details