Product Details
- SNP ID
-
rs1039341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:48540958 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGCAATCCTTCCTTCCCATAGC[C/T]GAGAAAGGGAAAGGGGACAGGCCAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
N4BP1
PubMed Links
Gene Details
- Gene
- N4BP1
- Gene Name
- NEDD4 binding protein 1
View Full Product Details