Product Details

SNP ID
rs3763965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27507440 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATTCTTATAAAATCTTAAGTAA[A/T]TGAAATATTGTGAAGCAAATAATAT
Phenotype
MIM: 611468 MIM: 612332
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BDNF-AS PubMed Links

Gene Details

Gene
BDNF-AS
Gene Name
BDNF antisense RNA
There are no transcripts associated with this gene.

Gene
LIN7C
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018362.3 Intron NP_060832.1
Gene
MIR8087
Gene Name
microRNA 8087
There are no transcripts associated with this gene.

View Full Product Details