Product Details

SNP ID
rs3733946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:133960201 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTACATTTCTTCTTAGTCAGTTAT[C/T]CTTCAGATAATAATTGCTTTGCTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf15 PubMed Links

Gene Details

Gene
C5orf15
Gene Name
chromosome 5 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020199.2 Intron NP_064584.1
Gene
LOC105379183
Gene Name
uncharacterized LOC105379183
There are no transcripts associated with this gene.

View Full Product Details