Product Details

SNP ID
rs11934921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99568321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAATTATAGTAAGGTCACATGAT[A/G]TAAGGTTAACATGCAAAAGTCAATT
Phenotype
MIM: 157147 MIM: 616013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTTP PubMed Links

Gene Details

Gene
MTTP
Gene Name
microsomal triglyceride transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000253.3 Intron NP_000244.2
NM_001300785.1 Intron NP_001287714.1
Gene
TRMT10A
Gene Name
tRNA methyltransferase 10A
There are no transcripts associated with this gene.

View Full Product Details