Product Details

SNP ID
rs10484775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150604252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGCTCTAATATTTGAAGTGGTG[A/T]ATAGATCTAGTTCTCTGGGTCTAGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHG1 PubMed Links

Gene Details

Gene
PLEKHG1
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005267064.3 Intron XP_005267121.1
XM_005267065.3 Intron XP_005267122.1
XM_005267066.2 Intron XP_005267123.1
XM_006715521.1 Intron XP_006715584.1
XM_006715522.3 Intron XP_006715585.1
XM_011535981.2 Intron XP_011534283.1
XM_011535982.2 Intron XP_011534284.1
XM_011535983.2 Intron XP_011534285.1
XM_017011102.1 Intron XP_016866591.1

View Full Product Details