Product Details

SNP ID
rs831592
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:33741647 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATATTTTAAAACAAAGGTATGTCC[A/G]TTCCTTAGCTAGAGAACTATTCTTC
Phenotype
MIM: 107271 MIM: 609089
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD59 PubMed Links

Gene Details

Gene
CD59
Gene Name
CD59 molecule
There are no transcripts associated with this gene.

Gene
FBXO3
Gene Name
F-box protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012175.3 1705 UTR 3 NP_036307.2
NM_033406.2 1705 Intron NP_208385.1
XM_011519980.1 1705 Intron XP_011518282.1
XM_011519981.2 1705 Intron XP_011518283.1
XM_011519982.1 1705 Intron XP_011518284.1
XM_017017533.1 1705 UTR 3 XP_016873022.1

View Full Product Details