Product Details

SNP ID
rs2419857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:113857617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATAGTTGTAATAGCTGCATTGA[A/G]TTATTTATAAGCAGTAAACTTTGTT
Phenotype
MIM: 609682
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCLRE1A PubMed Links

Gene Details

Gene
DCLRE1A
Gene Name
DNA cross-link repair 1A
There are no transcripts associated with this gene.

Gene
NHLRC2
Gene Name
NHL repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198514.3 Intron NP_940916.2
XM_011539769.2 Intron XP_011538071.1

View Full Product Details