Product Details

SNP ID
rs3807511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101235940 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTCCCCTCTCCTGCCTGTGTGA[C/T]TCAGGGTGGGGTGCTGCACCTTGCT
Phenotype
MIM: 615778 MIM: 609003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN15 PubMed Links

Gene Details

Gene
CLDN15
Gene Name
claudin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185080.1 Intron NP_001172009.1
NM_014343.2 Intron NP_055158.1
Gene
FIS1
Gene Name
fission, mitochondrial 1
There are no transcripts associated with this gene.

View Full Product Details