Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_005255867.1 | Intron | XP_005255924.1 | |||
XM_005255868.1 | Intron | XP_005255925.1 | |||
XM_011522970.1 | Intron | XP_011521272.1 | |||
XM_011522971.2 | Intron | XP_011521273.1 | |||
XM_011522973.2 | Intron | XP_011521275.1 | |||
XM_017023094.1 | Intron | XP_016878583.1 | |||
XM_017023095.1 | Intron | XP_016878584.1 | |||
XM_017023096.1 | Intron | XP_016878585.1 | |||
XM_017023097.1 | Intron | XP_016878586.1 | |||
XM_017023098.1 | Intron | XP_016878587.1 | |||
XM_017023099.1 | Intron | XP_016878588.1 | |||
XM_017023100.1 | Intron | XP_016878589.1 |