Product Details

SNP ID
rs12736534
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:108857312 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATTCCCTGTGTGATTTACTTTAT[C/T]ATGAAGTCTAAATTTGATGCTGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKNAD1 PubMed Links

Gene Details

Gene
AKNAD1
Gene Name
AKNA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152763.4 Intron NP_689976.2
Gene
SPATA42
Gene Name
spermatogenesis associated 42 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details