Product Details

SNP ID
rs12440410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58141487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGAATGGAAAAAAATGCAAGTAC[C/T]TCTGCTAAAAGTAGTACCCTTGATC
Phenotype
MIM: 602914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP9 PubMed Links

Gene Details

Gene
AQP9
Gene Name
aquaporin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320635.1 Intron NP_001307564.1
NM_001320636.1 Intron NP_001307565.1
NM_020980.4 Intron NP_066190.2
Gene
LOC101928635
Gene Name
uncharacterized LOC101928635
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011522293.2 Intron XP_011520595.1

View Full Product Details