Product Details
- SNP ID
-
rs2839196
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46288123 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACTGCTTCTGCAATATTTTTGCA[A/G]TTTTAAGTGGGATTCGTTAATGAAT
- Phenotype
-
MIM: 603294
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MCM3AP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78739486] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MCM3AP
- Gene Name
- minichromosome maintenance complex component 3 associated protein
There are no transcripts associated with this gene.
- Gene
- YBEY
- Gene Name
- ybeY metallopeptidase (putative)
View Full Product Details