Product Details

SNP ID
rs9465032
Assay Type
Functionally Tested
NCBI dbSNP Submissions
33
Location
Chr.6:17404947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGTATTGTATGAGTGTATATGTA[A/T]AACAGACAGACAGCACTACAGTATT
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CAP2 PubMed Links

Gene Details

Gene
CAP2
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006366.2 Intron NP_006357.1
XM_011514233.2 Intron XP_011512535.1

View Full Product Details