Product Details

SNP ID
rs1169312
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:121003658 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTCTCCTTTCTCAGACGATTCTGT[G/T]AAGTCTATGTGGGCTAAGGAGAGAA
Phenotype
MIM: 142410
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf43 PubMed Links

Gene Details

Gene
C12orf43
Gene Name
chromosome 12 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286191.1 1216 UTR 3 NP_001273120.1
NM_001286192.1 1216 UTR 3 NP_001273121.1
NM_001286195.1 1216 UTR 3 NP_001273124.1
NM_001286196.1 1216 UTR 3 NP_001273125.1
NM_001286197.1 1216 UTR 3 NP_001273126.1
NM_001286198.1 1216 UTR 3 NP_001273127.1
NM_022895.2 1216 UTR 3 NP_075046.1
XM_017019829.1 1216 Intron XP_016875318.1
Gene
HNF1A
Gene Name
HNF1 homeobox A
There are no transcripts associated with this gene.

View Full Product Details