Product Details

SNP ID
rs4973438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231523671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTCATCCTCCAGCTCCCAGCC[A/T]TGGAAGCCAGGGCTTCCTCTCAGGA
Phenotype
MIM: 604153
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00471 PubMed Links

Gene Details

Gene
LINC00471
Gene Name
long intergenic non-protein coding RNA 471
There are no transcripts associated with this gene.

Gene
NMUR1
Gene Name
neuromedin U receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006056.4 2751 UTR 3 NP_006047.3
XM_006712195.3 2751 Intron XP_006712258.1
XM_006712196.3 2751 Intron XP_006712259.1
XM_011510487.2 2751 UTR 3 XP_011508789.1
XM_011510488.2 2751 Intron XP_011508790.1
XM_011510489.2 2751 Intron XP_011508791.1

View Full Product Details