Product Details

SNP ID
rs2997922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128818465 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCGACGATGCCACGCCCCTCATAC[C/T]GGCGTCTTAAATGCGCAAAGAAAGG
Phenotype
MIM: 600440 MIM: 616218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENDOG PubMed Links

Gene Details

Gene
ENDOG
Gene Name
endonuclease G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004435.2 64 Intron NP_004426.2
XM_011518347.2 64 Silent Mutation ACC,ACT T16T XP_011516649.1
Gene
SPOUT1
Gene Name
SPOUT domain containing methyltransferase 1
There are no transcripts associated with this gene.

Gene
TBC1D13
Gene Name
TBC1 domain family member 13
There are no transcripts associated with this gene.

View Full Product Details