Product Details

SNP ID
rs301025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95517070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACTCACACTGTATTTTTTTTT[G/T]TGCAAGTTTAGATAGATAGCTTTTA
Phenotype
MIM: 601870 MIM: 610993
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
METAP2 PubMed Links

Gene Details

Gene
METAP2
Gene Name
methionyl aminopeptidase 2
There are no transcripts associated with this gene.

Gene
USP44
Gene Name
ubiquitin specific peptidase 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042403.2 3501 UTR 3 NP_001035862.1
NM_001278393.1 3501 UTR 3 NP_001265322.1
NM_032147.4 3501 UTR 3 NP_115523.2
XM_005269172.1 3501 UTR 3 XP_005269229.1
XM_005269173.1 3501 UTR 3 XP_005269230.1
XM_005269174.1 3501 UTR 3 XP_005269231.1
XM_011538800.2 3501 UTR 3 XP_011537102.1
XM_011538801.1 3501 UTR 3 XP_011537103.1
XM_011538802.1 3501 UTR 3 XP_011537104.1
XM_011538803.1 3501 UTR 3 XP_011537105.1
XM_011538804.2 3501 UTR 3 XP_011537106.1
XM_011538805.1 3501 UTR 3 XP_011537107.1
XM_011538806.2 3501 Intron XP_011537108.1
XM_017020009.1 3501 UTR 3 XP_016875498.1
XM_017020010.1 3501 UTR 3 XP_016875499.1
XM_017020011.1 3501 UTR 3 XP_016875500.1
XM_017020012.1 3501 Intron XP_016875501.1

View Full Product Details