Product Details
- SNP ID
-
rs7974458
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:95520750 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TAAGAAAAGTTAACACTCTGACTTT[C/T]AGTTAGCAACAAGACTATCCTCATT
- Phenotype
-
MIM: 601870
MIM: 610993
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
METAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77634106] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- METAP2
- Gene Name
- methionyl aminopeptidase 2
There are no transcripts associated with this gene.
- Gene
- USP44
- Gene Name
- ubiquitin specific peptidase 44
View Full Product Details