Product Details

SNP ID
rs2442803
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:12568575 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAGAGAACAGTTTTTCACAGAGT[A/G]TTTTCTCAATTATGTGGCTCACATT
Phenotype
MIM: 608426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKRN2 PubMed Links

Gene Details

Gene
MKRN2
Gene Name
makorin ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271707.1 Intron NP_001258636.1
NM_014160.4 Intron NP_054879.3
Gene
MKRN2OS
Gene Name
MKRN2 opposite strand
There are no transcripts associated with this gene.

View Full Product Details