Product Details

SNP ID
rs11741451
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40997357 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAGAAACTTGTTTTTAGGTAT[A/G]TAATTTCATGTATGTAACCTTTCAC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MROH2B PubMed Links

Gene Details

Gene
MROH2B
Gene Name
maestro heat like repeat family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173489.4 Intron NP_775760.3
XM_011513952.1 Intron XP_011512254.1
XM_011513953.1 Intron XP_011512255.1

View Full Product Details