Product Details

SNP ID
rs6128821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38726162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGCAGGCCGCAGCTTTCTGGGC[A/C]GAAGGAGACTTCAGCTCTAGGCAAC
Phenotype
MIM: 616440
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC32A1 PubMed Links

Gene Details

Gene
SLC32A1
Gene Name
solute carrier family 32 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080552.2 Intron NP_542119.1

View Full Product Details