Product Details

SNP ID
rs3750225
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:70055372 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCATGTGTTTGTTTAGGCTGGAAGA[C/T]TGAGAGAAACATTTCCCACATGTAG
Phenotype
MIM: 611781
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRDM14 PubMed Links

Gene Details

Gene
PRDM14
Gene Name
PR domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024504.3 1618 Silent Mutation CAA,CAG Q472Q NP_078780.1
XM_011517572.1 1618 Intron XP_011515874.1

View Full Product Details