Product Details

SNP ID
rs2105117
Assay Type
Functionally tested
NCBI dbSNP Submissions
71
Location
Chr.1:152511653 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGTCCCCCAAAATGCCCTCCCCAGT[G/A]TTCAGCCCCATGCCCACCTCCAGTC
Phenotype
MIM: 612619
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CRCT1 PubMed Links

Gene Details

Gene
CRCT1
Gene Name
cysteine rich C-terminal 1
There are no transcripts associated with this gene.

Gene
LCE5A
Gene Name
late cornified envelope 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178438.4 295 Missense Mutation TAT,TGT Y40C NP_848525.1

View Full Product Details